Assay Details
Target Gene Details
Entrez Gene ID: | 10528 |
Gene Name: | NOP56 ribonucleoprotein |
Gene Aliases: |
NOL5A, SCA36 |
Location: |
Chr.20:2652532-2658393 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NOP56 | NM_006392.3 | 6 | 788 | NP_006383.2 |
| NR_027700.2 | 6 | 788 | ||
| AK097186.1 | ||||
| AK292721.1 | 6 | 688 | ||
| BC018421.1 | 5 | 648 | ||
| BC104791.1 | 6 | 788 | AAI04792.1 | |
| BC104793.1 | 6 | 788 | AAI04794.1 | |
| BX647439.1 | 6 | 688 | ||
| BX647513.1 | 5 | 712 | ||
| Y12065.1 | 5 | 698 | CAA72789.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map