Assay Details
Target Gene Details
Entrez Gene ID: | 79576 |
Gene Name: | NFKB activating protein |
Gene Aliases: |
- |
Location: |
Chr.X:119925050-119943772 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAP | NM_024528.3 | 9 | 1529 | NP_078804.2 |
| XM_017029842.1 | 9 | 1066 | XP_016885331.1 | |
| AK026279.1 | 9 | 1529 | BAB15428.1 | |
| AK094446.1 | 7 | 2290 | ||
| AL832118.1 | 11 | 1470 | ||
| AY388958.1 | 9 | 1529 | AAQ90402.1 | |
| BC015354.1 | 9 | 1393 | AAH15354.1 | |
| BC032442.1 | 9 | 1387 | AAH32442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758888 | Chr.X:119823476 - 120195131 on Build GRCh38 | Gain |
|
| esv26416 | Chr.X:119900852 - 119931950 on Build GRCh38 | Gain |
|
| nsv7059 | Chr.X:119920556 - 119938506 on Build GRCh38 | Insertion |
|
| nsv9969 | Chr.X:119899907 - 119925214 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map