Assay Details
Target Gene Details
Entrez Gene ID: | 466 |
Gene Name: | activating transcription factor 1 |
Gene Aliases: |
EWS-ATF1, FUS/ATF-1, TREB36 |
Location: |
Chr.12:50763682-50821162 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATF1 | NM_005171.4 | 1 | 130 | NP_005162.1 |
| XM_011538387.2 | XP_011536689.1 | |||
| XM_011538388.2 | 1 | 50 | XP_011536690.1 | |
| XM_017019336.1 | XP_016874825.1 | |||
| AK313406.1 | 1 | 35 | ||
| BG545632.1 | 1 | 58 | ||
| BI712653.1 | ||||
| DA468394.1 | 1 | 130 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1069665 | Chr.12:50764016 - 50764617 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map