Assay Details
Target Gene Details
Entrez Gene ID: | 23613 |
Gene Name: | zinc finger MYND-type containing 8 |
Gene Aliases: |
PRKCBP1, PRO2893, RACK7 |
Location: |
Chr.20:47209214-47357780 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33497 | Chr.20:47143439 - 47650962 on Build GRCh38 | Loss |
|
| nsv1061899 | Chr.20:46498425 - 47439490 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map