Assay Details
Target Gene Details
Entrez Gene ID: | 10959 |
Gene Name: | transmembrane p24 trafficking protein 2 |
Gene Aliases: |
P24A, RNP24, p24, p24b1, p24beta1 |
Location: |
Chr.12:123584529-123598144 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED2 | NM_001321445.1 | NP_001308374.1 | ||
| NM_006815.3 | NP_006806.1 | |||
| AK298167.1 | ||||
| AK303189.1 | ||||
| AK312975.1 | ||||
| BC025957.1 | AAH25957.1 | |||
| BI086507.1 | ||||
| BP226125.1 | ||||
| CR541663.1 | CAG46464.1 | |||
| CR541682.1 | CAG46483.1 | |||
| DA559763.1 | ||||
| X92098.1 | CAA63069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422416 | Chr.12:123525129 - 123601142 on Build GRCh38 | Duplication |
|
| nsv1042181 | Chr.12:123552853 - 123668403 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map