Assay Details
Target Gene Details
Entrez Gene ID: | 200909 |
Gene Name: | 5-hydroxytryptamine receptor 3D |
Gene Aliases: |
5HT3D |
Location: |
Chr.3:184031544-184039369 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTR3D | NM_001145143.1 | NP_001138615.1 | ||
| NM_001163646.1 | NP_001157118.1 | |||
| NM_182537.2 | NP_872343.2 | |||
| XM_017005854.1 | XP_016861343.1 | |||
| AJ437318.1 | CAD24817.1 | |||
| AY159812.2 | ||||
| BC101090.2 | AAI01091.1 | |||
| BC101091.2 | AAI01092.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4145 | Chr.3:184023744 - 184056957 on Build GRCh38 | Insertion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77099580] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map