Assay Details
Target Gene Details
Entrez Gene ID: | 51291 |
Gene Name: | GEM interacting protein |
Gene Aliases: |
ARHGAP46 |
Location: |
Chr.19:19629473-19643667 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GMIP | NM_001288998.1 | NP_001275927.1 | ||
| NM_001288999.1 | NP_001275928.1 | |||
| NM_016573.3 | NP_057657.2 | |||
| XM_005259927.2 | XP_005259984.1 | |||
| XM_011528048.2 | XP_011526350.1 | |||
| XM_017026861.1 | XP_016882350.1 | |||
| XM_017026862.1 | XP_016882351.1 | |||
| AF132541.1 | AAF61330.1 | |||
| AK297220.1 | ||||
| BC126436.1 | ||||
| BC144142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952427 | Chr.19:19612592 - 19663391 on Build GRCh38 | Deletion |
|
| nsv1160593 | Chr.19:19623730 - 19685614 on Build GRCh38 | Deletion |
|
| dgv1025e212 | Chr.19:19629465 - 19639188 on Build GRCh38 | Loss |
|
| nsv509725 | Chr.19:19609994 - 19665351 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map