Assay Details
Target Gene Details
Entrez Gene ID: | 6494 |
Gene Name: | signal-induced proliferation-associated 1 |
Gene Aliases: |
SPA1 |
Location: |
Chr.11:65638107-65650930 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIPA1 | NM_006747.3 | 9 | 2292 | NP_006738.3 |
| NM_153253.29 | 9 | 2421 | NP_694985.29 | |
| XM_005274189.2 | 9 | 2366 | XP_005274246.1 | |
| XM_011545214.1 | 9 | 2332 | XP_011543516.1 | |
| AB005666.1 | 9 | 2421 | BAA22197.1 | |
| AF029789.1 | 9 | 2274 | AAC32547.1 | |
| AK303551.1 | 10 | 2060 | ||
| AK308234.1 | 9 | 2263 | ||
| AK310068.1 | ||||
| BC010492.1 | 9 | 2243 | AAH10492.1 | |
| BC110353.1 | 9 | 2261 | AAI10354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1969n54 | Chr.11:65568234 - 65663889 on Build GRCh38 | Loss |
|
| nsv951018 | Chr.11:65525430 - 65662029 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map