Assay Details
Target Gene Details
Entrez Gene ID: | 5662 |
Gene Name: | pleckstrin and Sec7 domain containing |
Gene Aliases: |
EFA6, EFA6A, PSD1, TYL |
Location: |
Chr.10:102402617-102420164 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSD | NM_001270965.1 | 16 | 2847 | NP_001257894.1 |
| NM_001270966.1 | 16 | 2368 | NP_001257895.1 | |
| NM_002779.4 | 15 | 3171 | NP_002770.3 | |
| NR_073110.1 | 6 | 943 | ||
| XM_011539968.1 | 16 | 2853 | XP_011538270.1 | |
| XM_011539969.2 | 16 | 2870 | XP_011538271.1 | |
| XM_017016433.1 | 16 | 2864 | XP_016871922.1 | |
| XM_017016434.1 | 15 | 2791 | XP_016871923.1 | |
| AB095931.1 | 15 | 3171 | ||
| BC039259.1 | 16 | 2355 | AAH39259.2 | |
| BC142643.1 | 16 | 2847 | ||
| BC142689.1 | 16 | 2847 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1159761 | Chr.10:102372778 - 102430809 on Build GRCh38 | Deletion |
|
| nsv525546 | Chr.10:102380593 - 102415138 on Build GRCh38 | Loss |
|
| nsv552058 | Chr.10:102397954 - 102410447 on Build GRCh38 | Loss |
|
| nsv951006 | Chr.10:102394644 - 102425743 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map