Assay Details
Target Gene Details
Entrez Gene ID: | 5813 |
Gene Name: | purine rich element binding protein A |
Gene Aliases: |
MRD31, PUR-ALPHA, PUR1, PURALPHA |
Location: |
Chr.5:140114123-140119416 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PURA | NM_005859.4 | 1 | 1218 | NP_005850.1 |
| BC036087.1 | 1 | 1171 | AAH36087.1 | |
| X91648.1 | 1 | 193 |
More Information
Set Membership: |
|

Genomic Map