Assay Details
Target Gene Details
Entrez Gene ID: | 23476 |
Gene Name: | bromodomain containing 4 |
Gene Aliases: |
CAP, HUNK1, HUNKI, MCAP |
Location: |
Chr.19:15236836-15332543 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRD4 | NM_058243.2 | 20 | 4555 | NP_490597.1 |
| XM_011527854.1 | 20 | 4620 | XP_011526156.1 | |
| AF386649.1 | 20 | 4555 | AAL26987.1 | |
| BC008354.1 | 4 | 787 | AAH08354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578717 | Chr.19:15159994 - 15241561 on Build GRCh38 | Loss |
|
| nsv953983 | Chr.19:15205390 - 15278689 on Build GRCh38 | Deletion |
|
| dgv3442n100 | Chr.19:15197257 - 15238422 on Build GRCh38 | Loss |
|
| esv34154 | Chr.19:15101623 - 15284812 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map