Assay Details
Target Gene Details
Entrez Gene ID: | 8079 |
Gene Name: | myeloid leukemia factor 2 |
Gene Aliases: |
NTN4 |
Location: |
Chr.12:6747992-6753470 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MLF2 | NM_005439.2 | 3 | 162 | NP_005430.1 |
| NR_026581.1 | 3 | 659 | ||
| AF070539.1 | 3 | 292 | AAC28632.1 | |
| AK289869.1 | 3 | 162 | ||
| AK313998.1 | 3 | 463 | ||
| BC000898.2 | 3 | 232 | AAH00898.1 | |
| BC002340.2 | 3 | 272 | AAH02340.1 | |
| BI544309.1 | ||||
| BT019393.1 | 2 | 99 | AAV38200.1 | |
| BT019432.1 | 2 | 99 | AAV38239.1 | |
| CR541700.1 | 2 | 99 | CAG46501.1 | |
| U57342.1 | 3 | 278 | AAB08988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3580394 | Chr.12:6696040 - 6779214 on Build GRCh38 | Gain |
|
| esv2761725 | Chr.12:6682182 - 6754772 on Build GRCh38 | Gain |
|
| nsv832322 | Chr.12:6575884 - 6765914 on Build GRCh38 | Loss |
|
| nsv832323 | Chr.12:6662233 - 6856559 on Build GRCh38 | Loss |
|
| nsv557257 | Chr.12:6720306 - 6789294 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map