Assay Details
Target Gene Details
Entrez Gene ID: | 102724919 |
Gene Name: | uncharacterized LOC102724919 |
Gene Aliases: |
- |
Location: |
Chr.1:186435161-186470291 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724919 | NR_126002.1 | |||
| DB056828.1 |
Target Gene Details
Entrez Gene ID: | 5132 |
Gene Name: | phosducin |
Gene Aliases: |
MEKA, PHD, PhLOP, PhLP |
Location: |
Chr.1:186443566-186461108 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDC | NM_002597.4 | 4 | 382 | NP_002588.3 |
| NM_022576.3 | 3 | 344 | NP_072098.1 | |
| XM_011509603.2 | 3 | 1400 | XP_011507905.1 | |
| AF076464.1 | 3 | 345 | AAD43142.1 | |
| BC093839.1 | 2 | 175 | AAH93839.1 | |
| BC112134.1 | 2 | 175 | AAI12135.1 | |
| EL945964.1 | ||||
| M33478.1 | 4 | 359 | AAA35486.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012164 | Chr.1:186354735 - 186596863 on Build GRCh38 | Gain |
|
| esv2762201 | Chr.1:186354747 - 186596875 on Build GRCh38 | Gain |
|
| esv3575361 | Chr.1:186266048 - 186604612 on Build GRCh38 | Gain |
|
| esv3890826 | Chr.1:185730915 - 186448352 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map