Assay Details
Target Gene Details
Entrez Gene ID: | 145553 |
Gene Name: | magnesium dependent phosphatase 1 |
Gene Aliases: |
FN6PASE, MDP-1 |
Location: |
Chr.14:24213937-24216070 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDP1 | NM_001199821.1 | NP_001186750.1 | ||
| NM_001199822.1 | NP_001186751.1 | |||
| NM_138476.3 | NP_612485.2 | |||
| AA918790.1 | ||||
| AK092821.1 | 4 | 479 | BAC03984.1 | |
| AK312038.1 | ||||
| BC046912.1 | AAH46912.1 | |||
| BC051382.1 | AAH51382.1 | |||
| CB046258.1 | ||||
| DA287183.1 |
More Information
Set Membership: |
|

Genomic Map