Assay Details
Target Gene Details
Entrez Gene ID: | 9839 |
Gene Name: | zinc finger E-box binding homeobox 2 |
Gene Aliases: |
HSPC082, SIP-1, SIP1, SMADIP1, ZFHX1B |
Location: |
Chr.2:144384375-144520391 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZEB2 | NM_001171653.1 | 1 | 249 | NP_001165124.1 |
| NM_014795.3 | 1 | 249 | NP_055610.1 | |
| NR_033258.1 | 1 | 249 | ||
| XM_011512232.2 | XP_011510534.1 | |||
| AB011141.1 | 1 | 171 | ||
| AB056507.1 | 1 | 207 | BAB40819.1 | |
| AK294928.1 | ||||
| AL118674.1 | 1 | 205 | ||
| BP218652.1 | 1 | 249 | ||
| DA305985.1 | 1 | 207 |
Target Gene Details
Entrez Gene ID: | 100303491 |
Gene Name: | ZEB2 antisense RNA 1 |
Gene Aliases: |
ZEB2-AS, ZEB2AS, ZEB2NAT |
Location: |
Chr.2:144519614-144520899 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZEB2-AS1 | NR_040248.2 | |||
| AI760227.1 | ||||
| BE048858.1 | ||||
| BG054540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3592768 | Chr.2:144379711 - 144546411 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map