Assay Details
Target Gene Details
Entrez Gene ID: | 64925 |
Gene Name: | coiled-coil domain containing 71 |
Gene Aliases: |
- |
Location: |
Chr.3:49162535-49166352 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC71 | NM_022903.3 | 2 | 1592 | NP_075054.3 |
| AK022862.1 | 2 | 1563 | BAB14278.1 | |
| AK023691.1 | 1 | 847 | BAB14642.1 | |
| AK098658.1 | 3 | 1427 | ||
| BC035516.1 | 2 | 1561 | AAH35516.1 | |
| BC071960.1 | 1 | 751 | AAH71960.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|

Genomic Map