Assay Details
Target Gene Details
Entrez Gene ID: | 9183 |
Gene Name: | zw10 kinetochore protein |
Gene Aliases: |
HZW10, KNTC1AP |
Location: |
Chr.11:113733183-113773763 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZW10 | NM_004724.3 | 12 | 1771 | NP_004715.1 |
| XM_017018558.1 | 11 | 1519 | XP_016874047.1 | |
| XM_017018559.1 | 12 | 1727 | XP_016874048.1 | |
| AK300746.1 | ||||
| AK303871.1 | 11 | 1450 | ||
| AK312925.1 | 12 | 1749 | ||
| BC128264.1 | 12 | 1699 | ||
| U54996.1 | 12 | 1697 | AAB88237.1 |
More Information
Set Membership: |
|

Genomic Map