Assay Details
Target Gene Details
Entrez Gene ID: | 29114 |
Gene Name: | transgelin 3 |
Gene Aliases: |
NP22, NP24, NP25 |
Location: |
Chr.3:111998739-112013888 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TAGLN3 | NM_001008272.1 | 5 | 1139 | NP_001008273.1 |
| NM_001008273.1 | 4 | 1024 | NP_001008274.1 | |
| NM_013259.2 | 5 | 1313 | NP_037391.2 | |
| AF087855.1 | 5 | 1025 | AAP97165.1 | |
| AF112201.1 | 4 | 810 | AAF17189.1 | |
| AF303058.1 | 5 | 1178 | AAL09330.1 | |
| AK315289.1 | 5 | 997 | ||
| BC015329.1 | 5 | 895 | AAH15329.1 | |
| BI822856.1 | ||||
| BM894229.1 | 1 | 191 |
More Information
Set Membership: |
|

Genomic Map