Assay Details
Target Gene Details
Entrez Gene ID: | 150465 |
Gene Name: | tubulin tyrosine ligase |
Gene Aliases: |
- |
Location: |
Chr.2:112482127-112532646 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTL | NM_153712.4 | 1 | 197 | NP_714923.1 |
| XM_005263599.3 | 1 | 236 | XP_005263656.1 | |
| XM_011510665.2 | 1 | 236 | XP_011508967.1 | |
| AB071393.2 | 1 | 42 | BAC06832.2 | |
| BC036819.1 | 1 | 209 | AAH36819.1 |
More Information
Set Membership: |
|

Genomic Map