Assay Details
Target Gene Details
Entrez Gene ID: | 886 |
Gene Name: | cholecystokinin A receptor |
Gene Aliases: |
CCK-A, CCK1-R, CCK1R, CCKRA |
Location: |
Chr.4:26481396-26490420 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCKAR | NM_000730.2 | NP_000721.1 | ||
| AK313978.1 | ||||
| L19315.1 | AAA02819.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762417 | Chr.4:26490146 - 26575178 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map