Assay Details
Target Gene Details
Entrez Gene ID: | 57149 |
Gene Name: | LYR motif containing 1 |
Gene Aliases: |
A211C6.1 |
Location: |
Chr.16:20899626-20925006 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436835 | Chr.16:18930652 - 21500976 on Build GRCh38 | Insertion |
|

Genomic Map