Assay Details
Target Gene Details
Entrez Gene ID: | 83875 |
Gene Name: | beta-carotene oxygenase 2 |
Gene Aliases: |
B-DIOX-II, BCDO2 |
Location: |
Chr.11:112175485-112218929 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCO2 | NM_001037290.2 | NP_001032367.2 | ||
| NM_001256397.1 | NP_001243326.1 | |||
| NM_001256398.1 | NP_001243327.1 | |||
| NM_001256400.1 | NP_001243329.1 | |||
| NM_031938.5 | NP_114144.4 | |||
| AB096252.1 | ||||
| AF276432.1 | AAK69433.1 | |||
| AJ290393.1 | CAC27994.1 | |||
| AK027801.1 | BAB55379.1 | |||
| AK091783.1 | BAC03747.1 | |||
| AK297853.1 | ||||
| AK315300.1 | ||||
| BC041656.1 | ||||
| BX503569.1 | ||||
| CD708042.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525636 | Chr.11:112172891 - 112192531 on Build GRCh38 | Loss |
|
| esv2761708 | Chr.11:112176838 - 112212857 on Build GRCh38 | Loss |
|
| esv2745077 | Chr.11:111949712 - 112188677 on Build GRCh38 | Deletion |
|
| nsv519192 | Chr.11:112160901 - 112193416 on Build GRCh38 | Gain |
|
| esv3579908 | Chr.11:112173319 - 112217076 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map