Assay Details
Target Gene Details
Entrez Gene ID: | 57731 |
Gene Name: | spectrin beta, non-erythrocytic 4 |
Gene Aliases: |
QV, SPNB4, SPTBN3 |
Location: |
Chr.19:40467153-40576459 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPTBN4 | NM_020971.2 | 7 | 807 | NP_066022.2 |
| XM_011527173.2 | 7 | 873 | XP_011525475.1 | |
| XM_017027049.1 | 7 | 1060 | XP_016882538.1 | |
| XM_017027050.1 | 5 | 442 | XP_016882539.1 | |
| XM_017027051.1 | 7 | 873 | XP_016882540.1 | |
| XM_017027052.1 | 7 | 873 | XP_016882541.1 | |
| AF082075.1 | 7 | 883 | AAG38874.1 | |
| AF311855.1 | 7 | 807 | AAG42473.1 | |
| AF324063.1 | 7 | 791 | AAQ14860.1 | |
| AF324064.1 | 7 | 791 | AAQ14859.1 | |
| AY004226.1 | 6 | 705 | AAF93171.1 | |
| AY004227.1 | 6 | 705 | AAF93173.1 |
More Information
Set Membership: |
|

Genomic Map