Assay Details
Target Gene Details
Entrez Gene ID: | 200172 |
Gene Name: | schlafen like 1 |
Gene Aliases: |
- |
Location: |
Chr.1:41015597-41023444 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFNL1 | NM_001168247.2 | 4 | 1157 | NP_001161719.1 |
| NM_001300859.1 | 4 | 887 | NP_001287788.1 | |
| NM_144990.3 | 4 | 1001 | NP_659427.3 | |
| XM_005270597.2 | 3 | 1740 | XP_005270654.1 | |
| XM_005270598.2 | 3 | 1056 | XP_005270655.1 | |
| XM_005270599.2 | 4 | 1064 | XP_005270656.1 | |
| XM_006710432.3 | 3 | 2151 | XP_006710495.1 | |
| XM_011540943.1 | 3 | 1687 | XP_011539245.1 | |
| XM_011540945.1 | 3 | 1624 | XP_011539247.1 | |
| XM_011540947.1 | 3 | 1149 | XP_011539249.1 | |
| XM_011540948.1 | 4 | 1602 | XP_011539250.1 | |
| XM_011540949.1 | 3 | 1532 | XP_011539251.1 | |
| XM_011540950.1 | 3 | 1502 | XP_011539252.1 | |
| XM_011540951.1 | 4 | 1056 | XP_011539253.1 | |
| XM_011540952.2 | 3 | 1479 | XP_011539254.1 | |
| XM_011540953.2 | 3 | 2059 | XP_011539255.1 | |
| XM_011540954.1 | 3 | 1410 | XP_011539256.1 | |
| XM_017000573.1 | 3 | 1740 | XP_016856062.1 | |
| XM_017000574.1 | 3 | 1560 | XP_016856063.1 | |
| AK097609.1 | 4 | 875 | ||
| AK292296.1 | 4 | 1127 | ||
| BC022037.2 | 4 | 963 | AAH22037.2 | |
| BC037879.1 | 2 | 1813 | AAH37879.1 | |
| BC050353.1 | 2 | 3321 | AAH50353.1 |
Target Gene Details
Entrez Gene ID: | 100507178 |
Gene Name: | SLFNL1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:41014590-41043890 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFNL1-AS1 | NR_037868.1 | 1 | 3260 | |
| AL157461.1 | 1 | 3260 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546103 | Chr.1:41011772 - 41076832 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map