Assay Details
Target Gene Details
Entrez Gene ID: | 3420 |
Gene Name: | isocitrate dehydrogenase 3 (NAD(+)) beta |
Gene Aliases: |
RP46 |
Location: |
Chr.20:2658390-2664223 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IDH3B | NM_001258384.2 | NP_001245313.1 | ||
| NM_006899.4 | NP_008830.2 | |||
| NM_174855.3 | NP_777280.1 | |||
| NR_136344.1 | ||||
| XM_005260716.1 | XP_005260773.1 | |||
| AK001905.1 | BAA91971.1 | |||
| AK300485.1 | ||||
| AK309488.1 | ||||
| AK315641.1 | ||||
| BC001960.1 | AAH01960.1 | |||
| BP872064.1 | ||||
| BX647393.1 | ||||
| HY131986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057464 | Chr.20:2650623 - 2989686 on Build GRCh38 | Gain |
|
| nsv470535 | Chr.20:1948896 - 2780594 on Build GRCh38 | Gain |
|
| esv3644998 | Chr.20:2148722 - 2690694 on Build GRCh38 | Gain |
|
| nsv458852 | Chr.20:2657933 - 2704278 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map