Assay Details
Target Gene Details
Entrez Gene ID: | 162979 |
Gene Name: | zinc finger protein 296 |
Gene Aliases: |
ZFP296, ZNF342 |
Location: |
Chr.19:45071500-45076430 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF296 | NM_145288.1 | 3 | 1432 | NP_660331.1 |
| AF447583.1 | 4 | 1478 | AAN76720.1 | |
| AY040679.1 | 2 | 1028 | AAK84667.1 | |
| BC019352.1 | 3 | 1432 | AAH19352.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953587 | Chr.19:45039143 - 45166142 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map