Assay Details
Target Gene Details
Entrez Gene ID: | 5926 |
Gene Name: | AT-rich interaction domain 4A |
Gene Aliases: |
RBBP-1, RBBP1, RBP-1, RBP1 |
Location: |
Chr.14:58298385-58373733 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARID4A | NM_002892.3 | NP_002883.3 | ||
| NM_023000.2 | NP_075376.2 | |||
| NM_023001.2 | NP_075377.2 | |||
| XM_005267964.2 | 1 | 137 | XP_005268021.1 | |
| XM_005267965.2 | 1 | 137 | XP_005268022.1 | |
| XM_017021560.1 | 1 | 137 | XP_016877049.1 | |
| XM_017021562.1 | 1 | 137 | XP_016877051.1 | |
| XM_017021563.1 | 1 | 137 | XP_016877052.1 | |
| AK307520.1 | ||||
| BC026230.1 | ||||
| DB218466.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv26881 | Chr.14:58298056 - 58298529 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map