Assay Details
Target Gene Details
Entrez Gene ID: | 393 |
Gene Name: | Rho GTPase activating protein 4 |
Gene Aliases: |
C1, RGC1, RhoGAP4, SrGAP4, p115 |
Location: |
Chr.X:153907376-153926260 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP4 | NM_001164741.1 | 23 | 3106 | NP_001158213.1 |
| NM_001666.4 | 22 | 2986 | NP_001657.3 | |
| AI249937.1 | 1 | 300 | ||
| AK294562.1 | 22 | 2911 | ||
| AY183123.1 | 19 | 2552 | AAO66337.1 | |
| BC023626.2 | 8 | 1160 | AAH23626.1 | |
| BC052303.1 | 23 | 3106 | AAH52303.1 | |
| D50921.1 | 22 | 2917 | BAA09480.1 | |
| X78817.1 | 22 | 2970 | CAA55394.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| nsv469867 | Chr.X:153813281 - 154018926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map