Assay Details
Target Gene Details
Entrez Gene ID: | 54974 |
Gene Name: | tRNA-histidine guanylyltransferase 1 like |
Gene Aliases: |
ICF45, IHG-1, hTHG1 |
Location: |
Chr.5:157731389-157741449 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| THG1L | NM_001317824.1 | 6 | 1056 | NP_001304753.1 |
| NM_001317825.1 | 6 | 1147 | NP_001304754.1 | |
| NM_001317826.1 | 7 | 1119 | NP_001304755.1 | |
| NM_017872.4 | 6 | 1151 | NP_060342.2 | |
| XM_017009603.1 | 6 | 1086 | XP_016865092.1 | |
| AF086486.1 | 1 | 327 | ||
| AK000553.1 | 6 | 1127 | BAA91249.1 | |
| AK021663.1 | 6 | 1102 | BAB13870.1 | |
| AK023355.1 | 6 | 724 | BAB14540.1 | |
| AK223119.1 | 6 | 1120 | BAD96839.1 | |
| AL136669.1 | 6 | 1119 | CAB66604.1 | |
| AY463216.1 | 6 | 1225 | AAS21134.1 | |
| BC001523.2 | 6 | 1097 | AAH01523.2 | |
| BC001852.2 | 6 | 1097 | AAH01852.2 | |
| BC023521.2 | 6 | 1117 | AAH23521.1 | |
| CR533503.1 | CAG38534.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759389 | Chr.5:157570600 - 157806036 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map