Assay Details
Target Gene Details
Entrez Gene ID: | 219968 |
Gene Name: | olfactory receptor family 5 subfamily B member 21 |
Gene Aliases: |
- |
Location: |
Chr.11:58507176-58508105 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR5B21 | NM_001005218.1 | 1 | 154 | NP_001005218.1 |
More Information
Set Membership: |
|

Genomic Map