Assay Details
Target Gene Details
Entrez Gene ID: | 23271 |
Gene Name: | calmodulin regulated spectrin associated protein family member 2 |
Gene Aliases: |
CAMSAP1L1 |
Location: |
Chr.1:200738905-200860707 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMSAP2 | NM_001297707.1 | 1 | 237 | NP_001284636.1 |
| NM_001297708.1 | 1 | 237 | NP_001284637.1 | |
| NM_203459.2 | 1 | 237 | NP_982284.1 | |
| XM_005245041.3 | 1 | 94 | XP_005245098.1 | |
| XM_017000799.1 | 1 | 890 | XP_016856288.1 | |
| AK095640.1 | 1 | 237 | ||
| BC125229.1 | ||||
| BC125230.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575438 | Chr.1:200664356 - 200857155 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map