Assay Details
Target Gene Details
Entrez Gene ID: | 221409 |
Gene Name: | spermatogenesis associated serine rich 1 |
Gene Aliases: |
DDIP, SPATA8, SRSP1 |
Location: |
Chr.6:44342660-44377167 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPATS1 | NM_145026.3 | NP_659463.1 | ||
| AA400657.1 | ||||
| AK058171.1 | BAB71701.1 | |||
| BC100956.2 | AAI00957.1 | |||
| BC100957.2 | AAI00958.1 | |||
| BC100958.2 | AAI00959.1 | |||
| BC100959.2 | AAI00960.1 | |||
| BC110065.1 | AAI10066.1 | |||
| BC111414.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3608829 | Chr.6:44369765 - 44371175 on Build GRCh38 | Loss |
|
| esv3567728 | Chr.6:44369759 - 44371197 on Build GRCh38 | Deletion |
|
| nsv1019145 | Chr.6:44345839 - 44453703 on Build GRCh38 | Gain |
|

Genomic Map