Assay Details
Target Gene Details
Entrez Gene ID: | 10042 |
Gene Name: | HMG-box containing 4 |
Gene Aliases: |
HMG2L1, HMGBCG, THC211630 |
Location: |
Chr.22:35257274-35295807 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMGXB4 | NM_001003681.2 | 5 | 941 | NP_001003681.1 |
| NR_027780.1 | 6 | 1056 | ||
| XM_006724100.3 | 4 | 1020 | XP_006724163.1 | |
| XM_006724101.3 | 4 | 1020 | XP_006724164.1 | |
| XM_006724102.1 | 5 | 924 | XP_006724165.1 | |
| AJ010070.1 | 6 | 1005 | CAA08992.1 | |
| AK299833.1 | 6 | 1021 | ||
| AL079310.1 | 6 | 1005 | CAB45240.1 | |
| BC004194.2 | 3 | 656 | AAH04194.1 | |
| BC017326.2 | 5 | 893 | AAH17326.1 | |
| BC025303.2 | 4 | 836 | AAH25303.1 | |
| BC052624.1 | 5 | 895 | AAH52624.1 | |
| BC062413.1 | 5 | 893 | AAH62413.1 | |
| BC104754.1 | 1 | 503 | AAI04755.1 | |
| BC104756.1 | 1 | 503 | AAI04757.1 | |
| BC111373.1 | 4 | 845 | AAI11374.1 | |
| CR456504.1 | 4 | 797 | CAG30390.1 |
More Information
Set Membership: |
|

Genomic Map