Assay Details
Target Gene Details
Entrez Gene ID: | 23144 |
Gene Name: | zinc finger CCCH-type containing 3 |
Gene Aliases: |
ZC3HDC3 |
Location: |
Chr.8:143437655-143549729 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZC3H3 | NM_015117.2 | 10 | 2438 | NP_055932.2 |
| XM_006716536.3 | 12 | 2827 | XP_006716599.2 | |
| XM_017013248.1 | 11 | 3458 | XP_016868737.1 | |
| XM_017013249.1 | 11 | 3000 | XP_016868738.1 | |
| BC038670.1 | 10 | 2421 | AAH38670.1 | |
| D63484.1 | 10 | 2398 | BAA09771.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470247 | Chr.8:143283403 - 143615483 on Build GRCh38 | Loss |
|
| nsv950012 | Chr.8:143358831 - 143442430 on Build GRCh38 | Deletion |
|
| nsv831487 | Chr.8:143434577 - 143520589 on Build GRCh38 | Gain |
|
| nsv612769 | Chr.8:143429115 - 143459926 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map