Assay Details
Target Gene Details
Entrez Gene ID: | 27112 |
Gene Name: | family with sequence similarity 155 member B |
Gene Aliases: |
CXorf63, TED, TMEM28, bB57D9.1 |
Location: |
Chr.X:69505204-69532508 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM155B | NM_015686.2 | 1 | 194 | NP_056501.2 |
| XM_011530908.2 | 1 | 225 | XP_011529210.1 | |
| XM_011530909.2 | 1 | 225 | XP_011529211.1 | |
| AF087142.1 | 1 | 193 | AAC62086.1 | |
| BC136518.1 | 1 | 181 | ||
| BC136519.1 | 1 | 181 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25466 | Chr.X:69503642 - 69506748 on Build GRCh38 | Loss |
|

Genomic Map