Assay Details
Target Gene Details
Entrez Gene ID: | 223 |
Gene Name: | aldehyde dehydrogenase 9 family member A1 |
Gene Aliases: |
ALDH4, ALDH7, ALDH9, E3, TMABADH |
Location: |
Chr.1:165662212-165698663 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH9A1 | NM_000696.3 | NP_000687.3 | ||
| XM_011509294.2 | XP_011507596.1 | |||
| AF172093.1 | AAF43600.1 | |||
| AK293520.1 | ||||
| AK301767.1 | ||||
| AK302183.1 | ||||
| AK302191.1 | ||||
| AK302654.1 | ||||
| AK312751.1 | ||||
| BC151140.1 | ||||
| BC151141.1 | ||||
| U34252.1 | AAB18827.1 | |||
| U50203.1 | AAB06721.1 | |||
| X75425.1 | CAA53176.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3587809 | Chr.1:165678149 - 165687881 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map