Assay Details
Target Gene Details
Entrez Gene ID: | 348094 |
Gene Name: | ankyrin repeat and death domain containing 1A |
Gene Aliases: |
- |
Location: |
Chr.15:64911902-64958700 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ANKDD1A | NM_182703.5 | NP_874362.3 | ||
| AK075298.1 | BAC11532.1 | |||
| AK098736.1 | BAC05399.1 | |||
| AK307698.1 | ||||
| BC027350.1 | AAH27350.1 | |||
| DR004550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833041 | Chr.15:64838459 - 64970380 on Build GRCh38 | Loss |
|
| nsv569703 | Chr.15:64756110 - 64930560 on Build GRCh38 | Loss |
|
| dgv31n68 | Chr.15:64706333 - 64929392 on Build GRCh38 | Loss |
|
| nsv1049654 | Chr.15:64843547 - 64960198 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map