Assay Details
Target Gene Details
Entrez Gene ID: | 26261 |
Gene Name: | F-box protein 24 |
Gene Aliases: |
FBX24 |
Location: |
Chr.7:100582490-100601117 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBXO24 | NM_033506.2 | 1 | 87 | NP_277041.1 |
| XM_017011961.1 | XP_016867450.1 | |||
| AK292240.1 | 1 | 94 | ||
| AK310487.1 | ||||
| AL136811.1 | CAB66745.1 | |||
| BC067293.1 | AAH67293.1 | |||
| BU656991.1 | 1 | 87 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
| nsv951386 | Chr.7:100566478 - 100682977 on Build GRCh38 | Deletion |
|
| nsv831072 | Chr.7:100564880 - 100765212 on Build GRCh38 | Loss |
|
| nsv5868 | Chr.7:100572375 - 100617023 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map