Assay Details
Target Gene Details
Entrez Gene ID: | 169522 |
Gene Name: | potassium voltage-gated channel modifier subfamily V member 2 |
Gene Aliases: |
KV11.1, Kv8.2, RCD3B |
Location: |
Chr.9:2717526-2730037 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNV2 | NM_133497.3 | 1 | 857 | NP_598004.1 |
| AF348983.1 | 1 | 647 | AAL83910.1 | |
| AK302215.1 | ||||
| BC101352.1 | 1 | 857 | AAI01353.1 | |
| BC101353.2 | 1 | 857 | AAI01354.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1030063 | Chr.9:2691667 - 2729595 on Build GRCh38 | Loss |
|
| nsv516425 | Chr.9:2690980 - 2790307 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map