Assay Details
Target Gene Details
Entrez Gene ID: | 55119 |
Gene Name: | pre-mRNA processing factor 38B |
Gene Aliases: |
NET1 |
Location: |
Chr.1:108692292-108701803 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF38B | NM_018061.2 | NP_060531.2 | ||
| NR_037185.1 | ||||
| XM_011541687.2 | XP_011539989.1 | |||
| XM_017001596.1 | XP_016857085.1 | |||
| XM_017001597.1 | XP_016857086.1 | |||
| XM_017001598.1 | XP_016857087.1 | |||
| AK001192.1 | BAA91546.1 | |||
| AL833950.1 | CAH10702.1 | |||
| BC007757.2 | AAH07757.1 | |||
| BC016296.1 | ||||
| BC024275.1 | ||||
| BC034581.1 | AAH34581.1 | |||
| BC040127.1 | AAH40127.1 | |||
| BC053838.1 | AAH53838.1 | |||
| BC107801.1 | AAI07802.1 | |||
| BC130346.1 | ||||
| BC132963.1 |
More Information
Set Membership: |
|

Genomic Map