Assay Details
Target Gene Details
Entrez Gene ID: | 259230 |
Gene Name: | sphingomyelin synthase 1 |
Gene Aliases: |
MOB, MOB1, SMS1, TMEM23, hmob33 |
Location: |
Chr.10:50305585-50623977 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SGMS1 | NM_147156.3 | 11 | 2234 | NP_671512.1 |
| XM_005269675.1 | 6 | 1926 | XP_005269732.1 | |
| XM_011539582.2 | 8 | 1838 | XP_011537884.1 | |
| XM_011539583.2 | 6 | 1715 | XP_011537885.1 | |
| AB154421.1 | 7 | 1617 | BAD16809.1 | |
| AK026683.1 | 7 | 1697 | ||
| AK296233.1 | 5 | 901 | ||
| AY280959.1 | 11 | 2200 | AAP37279.1 | |
| AY312431.1 | 5 | 1426 | AAQ82051.1 | |
| BC042899.1 | 5 | 1495 | AAH42899.1 | |
| Y14155.1 | 1 | 65 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521273 | Chr.10:50291885 - 50438087 on Build GRCh38 | Gain |
|
| esv3891809 | Chr.10:50242444 - 50647850 on Build GRCh38 | Gain |
|
| nsv1053293 | Chr.10:50222199 - 50342889 on Build GRCh38 | Gain |
|
| nsv1051087 | Chr.10:49977683 - 50687535 on Build GRCh38 | Loss |
|
| esv2762864 | Chr.10:50271185 - 50347655 on Build GRCh38 | Gain |
|
| dgv853n100 | Chr.10:50014227 - 50407682 on Build GRCh38 | Gain |
|
| esv3623195 | Chr.10:50270575 - 50358411 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map