Assay Details
Target Gene Details
Entrez Gene ID: | 8175 |
Gene Name: | splicing factor 3a subunit 2 |
Gene Aliases: |
PRP11, PRPF11, SAP62, SF3a66 |
Location: |
Chr.19:2236817-2248679 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SF3A2 | NM_007165.4 | 5 | 441 | NP_009096.2 |
| AK314815.1 | 5 | 434 | ||
| BC004434.1 | 5 | 411 | AAH04434.1 | |
| BC009903.2 | 5 | 434 | AAH09903.1 | |
| BC015804.1 | 5 | 394 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578382 | Chr.19:2194897 - 2260362 on Build GRCh38 | Loss |
|
| dgv6218n54 | Chr.19:2221793 - 2336009 on Build GRCh38 | Loss |
|
| nsv833706 | Chr.19:2131211 - 2249764 on Build GRCh38 | Loss |
|
| nsv578383 | Chr.19:2214058 - 2285197 on Build GRCh38 | Loss |
|
| nsv833707 | Chr.19:2187092 - 2372223 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116548172] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map