Assay Details
Target Gene Details
Entrez Gene ID: | 23080 |
Gene Name: | AVL9 cell migration associated |
Gene Aliases: |
KIAA0241 |
Location: |
Chr.7:32495426-32588741 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AVL9 | NM_015060.2 | NP_055875.1 | ||
| XM_005249668.2 | XP_005249725.1 | |||
| XM_005249669.2 | XP_005249726.1 | |||
| XM_005249670.2 | XP_005249727.1 | |||
| XM_005249671.4 | XP_005249728.1 | |||
| XM_011515227.2 | XP_011513529.1 | |||
| XM_017011891.1 | XP_016867380.1 | |||
| AK090619.1 | BAC03489.1 | |||
| AL833560.1 | CAH10387.1 | |||
| BC027724.2 | AAH27724.1 | |||
| DB477229.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map