Assay Details
Target Gene Details
Entrez Gene ID: | 8560 |
Gene Name: | delta 4-desaturase, sphingolipid 1 |
Gene Aliases: |
DEGS, DEGS-1, DES1, Des-1, FADS7, MIG15, MLD |
Location: |
Chr.1:224183208-224193441 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DEGS1 | NM_001321541.1 | 2 | 408 | NP_001308470.1 |
| NM_001321542.1 | 2 | 518 | NP_001308471.1 | |
| NM_003676.3 | 2 | 408 | NP_003667.1 | |
| XM_017002648.1 | 2 | 448 | XP_016858137.1 | |
| AF002668.1 | 2 | 350 | AAB62238.1 | |
| AF466375.1 | 2 | 359 | AAM12531.1 | |
| AK308232.1 | 2 | 376 | ||
| AU126996.1 | 2 | 408 | ||
| AY423730.1 | 2 | 356 | AAS00493.1 | |
| BC000961.2 | 2 | 390 | AAH00961.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832692 | Chr.1:224018834 - 224220015 on Build GRCh38 | Gain |
|
| nsv8857 | Chr.1:224172241 - 224195321 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map