Assay Details
Target Gene Details
Entrez Gene ID: | 6429 |
Gene Name: | serine and arginine rich splicing factor 4 |
Gene Aliases: |
SFRS4, SRP75 |
Location: |
Chr.1:29147738-29182125 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SRSF4 | NM_005626.4 | 6 | 1590 | NP_005617.2 |
| XM_011541951.2 | 5 | 1300 | XP_011540253.1 | |
| AB209856.1 | 8 | 7420 | BAD93093.1 | |
| AK223444.1 | BAD97164.1 | |||
| AK291509.1 | 6 | 1312 | ||
| AK293706.1 | 4 | 1058 | ||
| BC002781.2 | 6 | 1314 | AAH02781.1 | |
| BT007415.1 | 6 | 1217 | AAP36083.1 | |
| L14076.1 | 6 | 1264 | AAA36649.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3585602 | Chr.1:29138513 - 29154021 on Build GRCh38 | Gain |
|
| nsv509091 | Chr.1:29103220 - 29163899 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map