Assay Details
Target Gene Details
Entrez Gene ID: | 151636 |
Gene Name: | deltex E3 ubiquitin ligase 3L |
Gene Aliases: |
BBAP |
Location: |
Chr.3:122564338-122575203 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3L | NM_138287.3 | NP_612144.1 | ||
| AF484416.1 | AAL90859.1 | |||
| AK056406.1 | 1 | 587 | ||
| AK125086.1 | ||||
| AL833598.1 | ||||
| AY225123.1 | AAP57517.1 | |||
| AY780792.1 | AAV98362.1 | |||
| BC042191.1 | AAH42191.1 | |||
| BC060509.1 | AAH60509.1 | |||
| BX648267.1 | 1 | 3137 | ||
| BX648645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2672720 | Chr.3:122570146 - 122575337 on Build GRCh38 | Deletion |
|
| esv3597613 | Chr.3:122570126 - 122575297 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map