Assay Details
Target Gene Details
Entrez Gene ID: | 29850 |
Gene Name: | transient receptor potential cation channel subfamily M member 5 |
Gene Aliases: |
LTRPC5, MTR1 |
Location: |
Chr.11:2403962-2423424 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPM5 | NM_014555.3 | 17 | 2556 | NP_055370.1 |
| XM_017017628.1 | 18 | 2643 | XP_016873117.1 | |
| AF177473.1 | 17 | 2556 | AAF26288.1 | |
| AJ270996.1 | 17 | 2529 | CAB66342.1 | |
| BC093787.1 | 17 | 2553 | AAH93787.1 | |
| BC093789.1 | 17 | 2553 | AAH93789.1 | |
| BC143351.1 | 17 | 2553 | ||
| BC143353.1 | 17 | 2553 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951287 | Chr.11:2396771 - 2475670 on Build GRCh38 | Deletion |
|
| nsv469928 | Chr.11:2294174 - 2473462 on Build GRCh38 | Loss |
|
| nsv1037537 | Chr.11:2409367 - 2810311 on Build GRCh38 | Gain |
|
| nsv509382 | Chr.11:2376150 - 2439195 on Build GRCh38 | Insertion |
|
| nsv553047 | Chr.11:1733170 - 2418537 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map