Assay Details
Target Gene Details
Entrez Gene ID: | 6536 |
Gene Name: | solute carrier family 6 member 9 |
Gene Aliases: |
GLYT1 |
Location: |
Chr.1:43996483-44031492 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC6A9 | NM_001024845.2 | 10 | 1555 | NP_001020016.1 |
| NM_001261380.1 | 9 | 1502 | NP_001248309.1 | |
| NM_006934.3 | 9 | 1547 | NP_008865.2 | |
| NM_201649.3 | 10 | 1709 | NP_964012.2 | |
| NR_048548.1 | 9 | 1547 | ||
| XM_011542017.2 | 10 | 1528 | XP_011540319.1 | |
| XM_017002151.1 | 10 | 1463 | XP_016857640.1 | |
| XM_017002152.1 | 9 | 1437 | XP_016857641.1 | |
| XM_017002153.1 | 9 | 1208 | XP_016857642.1 | |
| AI884725.1 | ||||
| AK296438.1 | 9 | 1305 | ||
| AK298611.1 | 8 | 1160 | ||
| AK304057.1 | 10 | 1423 | ||
| AK315233.1 | 9 | 1550 | ||
| AK315933.1 | 9 | 1368 | ||
| S70609.1 | 10 | 1693 | AAB30784.1 | |
| S70612.1 | 9 | 1531 | AAB30785.1 |
More Information
Set Membership: |
|

Genomic Map