Assay Details
Target Gene Details
Entrez Gene ID: | 55234 |
Gene Name: | DNA replication regulator and spliceosomal factor |
Gene Aliases: |
BWD, SMU-1, fSAP57 |
Location: |
Chr.9:33041852-33076716 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMU1 | NM_018225.2 | 12 | 3469 | NP_060695.2 |
| XM_005251503.4 | 11 | 3305 | XP_005251560.1 | |
| AK001732.1 | 1 | 1538 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv614143 | Chr.9:32583509 - 33141754 on Build GRCh38 | Gain |
|
| dgv7515n100 | Chr.9:33042019 - 33241605 on Build GRCh38 | Gain |
|
| esv3620303 | Chr.9:32569144 - 33069475 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map