Assay Details
Target Gene Details
Entrez Gene ID: | 25999 |
Gene Name: | CAP-Gly domain containing linker protein 3 |
Gene Aliases: |
CLIPR-59, CLIPR59, RSNL1 |
Location: |
Chr.19:36014660-36032895 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLIP3 | NM_001199570.1 | 13 | 2694 | NP_001186499.1 |
| NM_015526.2 | 14 | 2610 | NP_056341.1 | |
| AJ427922.1 | 14 | 2588 | CAD20873.1 | |
| AK056445.1 | 12 | 2093 | ||
| AK092354.1 | 10 | 1857 | ||
| AK094738.1 | 14 | 2608 | ||
| AL117468.1 | 6 | 1222 | CAB55943.1 | |
| BC014486.1 | 11 | 2053 | AAH14486.2 |
Target Gene Details
Entrez Gene ID: | 101927572 |
Gene Name: | uncharacterized LOC101927572 |
Gene Aliases: |
- |
Location: |
Chr.19:36014499-36045997 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC101927572 | NM_001290056.1 | NP_001276985.1 | ||
| HY030864.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv1000212 | Chr.19:36003741 - 36017606 on Build GRCh38 | Deletion |
|
| dgv51n68 | Chr.19:35841011 - 36016692 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map